Mouse T-cell Surface Glycoprotein CD3 Epsilon Chain (CD3E/T3E) ELISA Kit
Catalog # 76700-042
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Mouse
- Target protein:T-cell surface glycoprotein CD3 epsilon chain
- Description:Mouse T-cell surface glycoprotein CD3 epsilon chain (CD3E/T3E) ELISA kit
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf life:6 months (4 °C); 12 months (−20 °C)
- Detection range:1.25 - 40 ng/ml
- Storage temperature:2…8 °C
- Application:Immunology
- Sample volume:50 - 100 μl
- Sensitivity:0.156 ng/ml
- Regulatory status:RUO
- Cat. no.:76700-042
Specifications
About this item
- Detection wavelength: 450 nm