60 Peg Wall Mounted Draining Rack
60 peg wall mounted draining rack
Catalog # 470345-458
Supplier: Wards
Supplier #: CH0715E
Jump to:
Product Details & Documents
With 60 pegs that are 3" long, this wall-mounted rack made of vinyl-coated steel provides extensive space for efficient drying of lab equipment.
- Peg drying rack featuring a sturdy vinyl-coated steel construction that will stand up to the wear and tear of the lab
- Supports up to 60 test tubes with 3" pegs - Provides convenient insertion and removal of tubes
- Stands upright or hangs on the wall (mounting hardware not included)
- Designed for use in professional laboratories, but is an ideal choice for science classrooms
The sturdy wall mounted laboratory drying rack contains 60 test tube pins. Each pin is 3" long and pegs are 2.5" apart. The rack is made from vinyl coated steel and each peg features a rubber cap to prevent damages and scratches to glassware. Accessories to mount the rack are not included.
Specifications
- Description:60 peg wall mounted draining rack