Order Entry
Canada
Orders LinkContactUsLinkComponent
Slotted Base

Slotted Base

Slotted base
Catalog # 470345-852
Supplier:   Wards
Supplier #:  CH0660
Slotted Base
Catalog # 470345-852
Supplier:  Wards
Slotted Base
Slotted Base

Slotted base

Price
72,05 $
Log in for availability
Each
Product Details & Documents

Slotted base made of chemical-resistant cast metal, offering durability and stability for laboratory apparatus and setups.

  • Cast metal slotted base
  • Chemical resistant
  • Accommodates objects up to 1.57" (40 mm)
  • Convenient thumbscrew for rapid tightening and release
  • 3.4 lbs

Cast metal slotted base is finished in a chemical resistant paint. Its jaws open to accommodate objects of up to 1.57". The dimensions of the base are 3×3.5" and the depth of the jaws is 1.75".

Specifications
  • Description:
    Slotted base
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.