ContactUsLinkComponent
7-Piece Filter Adapter Tapered Cones Set
7-Piece Filter Adapter Tapered Cones Set
Catalog # 470345-184
Supplier:  Wards
undefined
7-Piece Filter Adapter Tapered Cones Set
Catalog # 470345-184
Supplier:  Wards
Supplier Number:  CH0421
Specifications
  • Description:
    7-piece filter adapter tapered cones set
  • Environmentally Preferable:
  • Cat. no.:
    470345-184
Specifications
About this item
Set of 7 neoprene rubber tapered cones specifically designed for Buchner funnels, providing a secure and efficient filtration system, available in multiple sizes for versatile laboratory applications.
  • Size 1 to 7 tapered, blue-colored filter adapter cones
  • Designed for use with Buchner funnels in filtration assemblies
  • Made of quality, flexible neoprene rubber - Good chemical stability and can withstand a wide temperature range - Autoclavable
  • Can be nested within different size cones for convenient storage
  • Cones vary in size - Height: 20 to 43 mm, Top: 22 to 81 mm, Bottom: 11 to 57 mm
Set of 7 tapered filter adapter cones. These adapter cones are made for use with Buchner Funnels in filtration assemblies.