ContactUsLinkComponent
Hand Crank Centrifuge
Hand Crank Centrifuge
Catalog # 470344-568
Supplier:  Wards
undefined
Hand Crank Centrifuge
Catalog # 470344-568
Supplier:  Wards
Supplier Number:  CH0266B
Specifications
  • Description:
    Hand crank centrifuge
  • Environmentally Preferable:
  • Cat. no.:
    470344-568
Specifications
About this item
Hand crank centrifuge with a capacity of 4 tubes (up to 15 ml), includes tubes for immediate use.
  • An economic centrifuge that clamps right on to any bench top
  • Holds 4 tubes with 15 ml maximum capacity
  • Gear box contains grease and fittings for maintenance
  • Adjustment to crank and gearing will be needed once installed on table top to ensure easy use
Delivery: Supplied with table clamp, hand crank, 4 buckets to hold up to 15 ml tubes and 4 polythene tubes.