Order Entry
Canada
Orders LinkContactUsLinkComponent
Synthware High Vacuum Valve, PTFE, O-Ring, T-Type

Synthware High Vacuum Valve, PTFE, O-Ring, T-Type

High vacuum valve, PTFE, O-Ring, T-type, 0 - 12 mm
Catalog # 60004-012
Supplier:   Kemtech America
Supplier #:  S590012
Synthware High Vacuum Valve, PTFE, O-Ring, T-Type
Catalog # 60004-012
Supplier:  Kemtech America
Synthware High Vacuum Valve, PTFE, O-Ring, T-Type
Synthware High Vacuum Valve, PTFE, O-Ring, T-Type

High vacuum valve, PTFE, O-Ring, T-type, 0 - 12 mm

Price
$239.09
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

With two 9 mm O.D. sidearms sealed to the barrel at a 90° angle to one another.

  • Heavy wall construction
  • 33 expansion Borosilicate glass

The non-rotating PTFE plug has three O-Rings. The tip O-Ring makes the primary seal on the precision formed taper seat. For replacement Viton O-Ring kits, see R110 and R112.

Specifications
  • Description:
    High vacuum valve, PTFE, O-Ring, T-type
  • Valve size:
    0 - 12 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.