Orders LinkContactUsLinkComponent
Synthware Adapters with Ball Socket
Synthware Adapters with Ball Socket
Supplier:   Kemtech America
Synthware Adapters with Ball Socket
Supplier:  Kemtech America
Certificates
Synthware Adapters with Ball Socket
Synthware Adapters with Ball Socket
2 Options Available Below
Product Details & Documents
Ball socket adapter with lower standard taper inner joint.
  • Heavy wall construction
  • 33 expansion Borosilicate glass
  • Standard ground glass joints work with other brands
For use with any 35/20 ball socket joint.
Product Family Options
Product Information
  • Top [ST] joint size
    Bottom [ST] joint size
    Availability
    Price
  • 60000-090
    Adapter, with ball socket
    Adapter, with ball socket
    [ST] 35/20
    14/20
    Log in for availability
    Log in for availability
    Each
    Each - $70.72
    $70.72
    New Customer Promotional Price
    Each
       $70.72
    Minimum order quantity is 1
    Supplier Number: A663514
  • 60000-094
    Adapter, with ball socket
    Adapter, with ball socket
    [ST] 35/20
    24/20
    Log in for availability
    Log in for availability
    Each
    Each - $73.17
    $73.17
    New Customer Promotional Price
    Each
       $73.17
    Minimum order quantity is 1
    Supplier Number: A663524