About this item
Magid® D-ROC® GPD883 18-gauge work gloves features AeroDex® for the lightest cut resistance on the market and a VersaTek Grip™ palm coating for extreme durability, dexterity, and grip in any environment.
- AeroDex® is specially engineered to make hands feel cooler than a comparable cut resistant gloves
- Intelligent VersaTek Grip™ adapts to amplify grip ability in wet, dry, or oily environments
- Reinforced thumb saddle enhances durability at a critical wear point
- Touchscreen compatible to maintain user safety while operating electronic devices
- Shield graphic is color-coded by cut level to easily identify cut protection from a distance
- Machine washable for longer service life
- ANSI Level A8 Cut Resistance, ANSI Level 6 Abrasion Resistance, and ANSI Level 3 Puncture Resistance
VersaTek is a revolutionary coating technology engineered for maximum comfort and flexibility with stabilizers that enhance performance and durability. VersaTek Adaptive Coating Technology delivers unmatched durability while flexing with natural hand movements for precision handling of small parts. The GPD883 achieves an A8 cut level and is over 50% lighter in weight than comparable cut-resistant materials, all with a coating that reads the environment and adapts to what is needed to get the job done.
AeroDex® engineered yarn delivers extremely lightweight A8 cut resistance that is 50% lighter than a traditional HPPE of the same cut level. The 18-gauge AeroDex® shell delivers unmatched dexterity and tactile sensitivity making it agile enough for any job.
Specifications
- Catalog No:
- 76635-820
- 76635-822
- 76635-824
- 76635-810
- 76635-812
- 76635-814
- 76635-816
- 76635-818
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Gauge:
- 18
- 18
- 18
- 18
- 18
- 18
- 18
- 18