Azure Aqua Power Supply, 600 V
Catalog # 76414-794
Supplier: Azure Biosystems
Irregular Voltage
CAS Number:
Specifications
- Description:Azure Aqua power supply
- Cat. no.:76414-794
Specifications
About this item
The Azure Aqua Power Supply is a universal power supply that is designed for powering electrophoresis and Western blotting transfer modules. The power supply can create and save protocols for all your gel and blotting experiments, and includes a pause/resume function for flexibility.
- W×D×H: 10×18×10 cm
Power supply includes output range of 5 to 600 V, 1 to 1200 mA, 1 to 500 W, LCD readable voltage, current, power, and four power connections to run multiple experiments at once. Able to control constant power, current, or voltage.