ContactUsLinkComponent
Azure Aqua Transfer Cell
Azure Aqua Transfer Cell
Catalog # 76414-792
Supplier:  Azure Biosystems
CAS Number:  
undefined
Azure Aqua Transfer Cell
Catalog # 76414-792
Supplier:  Azure Biosystems
Supplier Number:  AC4201
CAS Number:  
Specifications
  • Description:
    Azure Aqua transfer cell
  • Cat. no.:
    76414-792
Specifications
About this item
The Azure Aqua transfer cell is used for transferring two mini gels to membranes for Western blotting experiments (wet transfer).
  • W×D×H: 10×18×10 cm
The transfer cassettes and electrode core are colored for directionality which makes transferring easy and straightforward. You can perform a quick one hour transfer or you can transfer overnight at a lower voltage.
Ordering information: Transfer cell includes 2 gel transfer cassette holders, 6 sponges, 2 Azure Aqua cooling units, electrode assembly, tank and top lid with power cables.