Order Entry
Canada
ContactUsLinkComponent
Azure Aqua Quad Mini-Cell
Azure Aqua Quad Mini-Cell
Catalog # 76414-790
Supplier:  Azure Biosystems
CAS Number:  
undefined
Azure Aqua Quad Mini-Cell
Catalog # 76414-790
Supplier:  Azure Biosystems
Supplier Number:  AC4200
CAS Number:  

Specifications

  • Capacity:
    Can fit up to 4 gels
  • Description:
    Azure Aqua Mini-cell tank, acrylic material, primarily used for nucleic acid and protein sequencing via vertical gel electrophoresis
  • Cat. no.:
    76414-790

Specifications

About this item

Compatible with mini gels cassette dimensions 10×8 cm.

  • Comes with 1 mm gel plate set
  • For use with polyacrylamide gels
  • Comprised of an upper and a lower buffer chamber

W×D×H: 14×10×15 cm

Ordering information: It includes 4 gel casting stands and casting frames, 1 buffer dam, 5 spacer plates (1mm), 10 short glass plates, 5 15-well combs, 5 10-well combs, 5 sealing gaskets, 5 gel shovels, Tank and upper lid with power cables, primary and secondary electrode cores.