ContactUsLinkComponent
Equipotential Trough Kit
Equipotential Trough Kit
Catalogue # 470349-678
undefined
Equipotential Trough Kit
Catalogue # 470349-678
Supplier Number:  UNEQPTK01
Spécifications
  • Description:
    Equipotential trough kit
  • Cat. no.:
    470349-678
Spécifications
À propos de cet article
Plot equipotential lines accurately with water and a digital multimeter.
  • Plot equipotential lines for various electrode configurations
  • Classic design
  • Correlates to NGSS Standards MS-PS2-5 and HS-PS2-4
  • Activity guide included
This comprehensive kit allows students to plot equipotential lines in an electric field between two electrodes held at different electric potentials. A simple setup keeps electrodes partially immersed in water containing an electrolyte, allowing a small current to flow between the electrodes. Students can then sample the resulting electric field using the included multimeter.