Green Laser Pointer
Green laser pointer, Class IIIa, 6L×0.5"Ø, 200 m (600') range
Catalog # 470320-710
Supplier: United Scientific Supplies
Supplier #: GLP006
Jump to:
Product Details & Documents
Bright, beautiful green laser pointer.
- Max. power 5 mW (Class 3R / IIIa)
- Approximate wavelength 532 nm
- Range 200 m (600')
- Presentation aid, and perfect for diffraction experiments
Economical laser pointers are useful for many optical experiments. Lasers operate on a push-button switch, are housed in a rubberized aluminum pen-style case, and include a pocket clip. The lasers are Class 3R.
Green laser pointer has a maximum output of 5mW and a wavelength of 532 nm (±10 nm). Range of beam is 200 meters. Expected operating life is 1000 hours.
: Operates on two 'AAA' batteries (not included).
Specifications
- Description:Green laser pointer, Class IIIa, 6L×0.5"Ø, 200 m (600') range



