Red Laser Pointer
Red laser pointer, Class IIIa, 6L×0.5"Ø, 100 m (300') range
Catalog # 470320-708
Supplier: United Scientific Supplies
Supplier #: RLP006
Jump to:
Product Details & Documents
Economical laser pointer, useful for many optical experiments.
- Max. power 5 mW (Class 3R / IIIa)
- Approximate wavelength 650 nm
- Range 100 m (300')
- Presentation aid, and perfect for diffraction experiments
Economical laser pointers are useful for many optical experiments. Lasers operate on a push-button switch, are housed in a rubberized aluminum pen-style case, and include a pocket clip. The lasers are Class 3R.
Red laser pointer has a maximum output of 5mW and a wavelength of 650 nm (±10 nm). Range of beam is 100 meters. Expected operating life is 1000 hours.
Ordering information: Operates on two 'AAA' batteries (not included).
Specifications
- Description:Red laser pointer, Class IIIa, 6L×0.5"Ø, 100 m (300') range



