Orders LinkContactUsLinkComponent
Slide Storage Boxes, Hinged, Plastic
Slide Storage Boxes, Hinged, Plastic
Slide Storage Boxes, Hinged, Plastic
Certificates
Slide Storage Boxes, Hinged, Plastic
Slide Storage Boxes, Hinged, Plastic
3 Options Available Below
Product Details & Documents
Store slides securely and safely.
  • Engraved numbers
  • Secure closing clasp
  • Key card for easy reference
Sturdy two-piece molded slide boxes hold standard 3×1" microscope slides. The hinged top has a reference card for labeling the corresponding slide description. The numbers on the opposite side are engraved for easy reference. These boxes are made from high impact polystyrene.
Product Family Options
Product Information
  • Material
    Capacity
    Availability
    Price
  • 470349-718
    Slide storage box, hinged
    Slide storage box, hinged
    Plastic
    25 slides
    Log in for availability
    Log in for availability
    Each
    Each - $7.25
    $7.25
    Each
       $7.25
    Supplier Number: 62211
  • 470349-770
    Slide storage box, hinged
    Slide storage box, hinged
    Plastic
    50 slides
    Log in for availability
    Log in for availability
    Each
    Each - $9.65
    $9.65
    Each
       $9.65
    Supplier Number: 62212
  • 470349-780
    Slide storage box, hinged
    Slide storage box, hinged
    Plastic
    100 slides
    Log in for availability
    Log in for availability
    Each
    Each - $16.25
    $16.25
    Each
       $16.25
    Supplier Number: 62213
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.