Lab-Tips® Small Knitted Polyester Swab with Flexible Tip
Small cleanroom swab with flexible tip, Polyester, 100/pack, 20 packs/case
Catalog # 89137-163
Supplier: Berkshire
Supplier #: LTP70F.20
Jump to:
Product Details & Documents
The LTP70F Swab is an excellent swab for cleaning grooved, slotted, and recessed areas.
- Cleanroom laundered, double layer, 100% polyester knit swab
- Critically low particles, fibers, ions, and extractables
- High abrasion resistance
- Chemically compatible with IPA, acetone, and other solvents
- Soft and non-abrasive with no contaminating adhesives
- Good sorbency
- Free from Silicone, amides, and DOP
- Autoclavable
The LTP70F Swab features a cleanroom laundered, knitted polyester head that is extremely clean and durable. A compact handle and internal flexible paddle provide maximum dexterity in confined areas.
Swabs are laundered and packaged in a Class 10 (ISO Class 4) cleanroom.
Specifications
- Description:Small cleanroom swab with flexible tip
- Material:Polyester
- Packaging:100/pack, 20 packs/case