ContactUsLinkComponent
pHep®4 pH and Temperature Tester, 0.1 pH Resolution
 :  Hanna
undefined
pHep®4 pH and Temperature Tester, 0.1 pH Resolution
 :  Hanna
  • Catalog No:
  • CA470004-986
  • CA470100-624
  • pH range:
  • –2...+16
  • pH resolution:
  • 0.1
  • pH accuracy:
  • ±0.1
  • pH calibration:
  • Automatic, one or two-point with two sets of standard buffers (pH 4.01/7.01/10.01 or pH 4.01/6.86/9.18)
Frequently Bought Together