Secador® 5.0 Desiccator Cabinets, Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
The ATS Life Sciences Bel-Art Secador® 5.0 Desiccator Cabinets provide dry storage for humidity-sensitive products such as laboratory samples, reagents, optical equipment, electronics, and 3D printing filament. Available in two configurations, a non-electric model that can be used with any Bel-Art desiccant and an electric model that is desiccant-free.
- High-density foam gaskets to keep ambient humidity out and dry air inside
- Large clear polycarbonate panels are durable and allow easy viewing of internal contents
- LCD shows the internal temperature and humidity level
- Includes three adjustable, removable shelves
- Desiccant free model keeps internal humidity at 20 to 40%
Ordering information: Supplied with three shelves, an additional shelf can be purchased separately.
Specifications
- Catalog No:
- 76638-964
- 76638-962
- Electrical:
- 100 - 240 VAC
- Non-electric
- Exterior dimensions:
- 30×38×63.5 cm
- 30×38×52 cm
- Int. volume:
- 45.3 L (1.6 cu.ft.)
- 45.3 L (1.6 cu.ft.)
Specifications
Customers Who Bought This Also Bought