Acro® 500 Filter Capsule with Supor® EKV Membrane and Polycarbonate Housing, Cytiva (Formerly Pall Lab)
Catalog # CA76640-756
Supplier: Avantor
Specifications
- Pore Size:0.65/0.2 µm
- Description:Acro® 500
- Cat. no.:CA76640-756
- Connection:12.7 mm (½") hose barb
- Effective Filtration Area:500 cm²
Specifications
About this item
The Acro 500 filter capsule with Supor EKV membrane and polycarbonate housing offers a 500 cm² effective filtration area (EFA) for the sterile filtration of larger volumes of laboratory solutions.
- Membrane features a built-in dual layer polyethersulfone (PES) construction which provides fast and efficient filtration
The low protein and preservative binding properties of the membrane allow for maximum transmission of active ingredients as well as broad liquid compatibility, ideal for the filtration of a wide range of fluids including buffers, biological fluids, and tissue culture media. With an outer housing made from polycarbonate, the improved optical clarity allows for easier venting of the capsule when in use to maximize membrane area and ultimately flow rate and throughput.
Frequently Bought Together