Order Entry
Canada
ContactUsLinkComponent
Mini Mixer, Electron Microscopy Sciences
Mini Mixer, Electron Microscopy Sciences
  100493-504
Minority or Woman-Owned Business Enterprise
undefined
Mini Mixer, Electron Microscopy Sciences
  100493-504
 :  71849-10
Minority or Woman-Owned Business Enterprise

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Description:
    Mini Mixer
  • Cat. no.:
    100493-504
  • Supplier No.:
    71849-10

 

 

Perfect for mixing in test tubes, micro centrifuge tubes and other containers which are too narrow for conventional mixers.

The stirring rod will also flare out for use in larger spaces.

Dimensions (including rod): 8 x 1/4 x 1"
Weight: 3 oz.

 : Comes with a clear vinyl carrying case, 4 1/4 x 1/4"stirring rod, and two "AA" batteries.

Replacement rods