Titanium(III) chloride solution 20% (w/w)
Catalog # CATX0700-2
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle, pre-packed
- Size:500 ml
- Cat. no.:CATX0700-2
- Supplier No.:TX0700-2
Specifications
About this item
:
:154.24
CAS Number:7705-07-9
Formula:Cl₃Ti
MDL Number:MFCD00011266