ContactUsLinkComponent
AlphaTec® EPDM Isolator Gloves, 85-505 Series, Ansell
AlphaTec® EPDM Isolator Gloves, 85-505 Series, Ansell
Supplier:  Ansell Healthcare
Certificates
undefined
AlphaTec® EPDM Isolator Gloves, 85-505 Series, Ansell
Supplier:  Ansell Healthcare
Specifications
  • Catalog No:
  • 76408-506
  • Model:
  • 85-505
  • Ambidextrous/Hand-specific:
  • Ambidextrous
  • Material:
  • EPDM
  • Finish:
  • Non-clean, fully dipped
  • Cuff style:
  • Beaded
  • Cuff thickness:
  • 24 mil
  • Palm thickness:
  • 24 mil