Electrical Breaker Lockout Tagout Kit with Nylon Safety Padlock in Pouch, Brady Worldwide
76637-582
- Color:Red
- Description:Lockout Kit, keyed different
- Dimensions:5H×7W×2"D
- Material:Nylon
- Cat. no.:76637-582
Safely power down faster when powering up lockout tagout readiness with this convenient, easily accessible and portable lockout kit.
- OSHA compliance driver
- Built to last
- Extremely portable
- Great for contractors
- Stores easily in vehicles to travel between jobsites
Designed and assembled exclusively for safety to help users comply with OSHA 29 CFR § 1910.147 Control of Hazardous Energy. All components meet Brady’s highest standards for quality, durability and reliability.
Belt-attachable pouch keeps tools at the ready and goes anywhere. All components are specially designed for electrical lockout scenarios, including non-conductive nylon-body padlock. Included lockout devices cover 120/277 V and 480/600 V breakers in single- and multi-pole configurations.
: Lockout Kit includes: (1) Nylon Safety Lockout Padlock, (1) Lockout Pouch, (1) 1" Lockout Hasp, (2) Clamp-on Circuit Breaker Lockouts 120/277 V, (1) Clamp-on Circuit Breaker Lockout 480/600 V, (5) Lockout Tags, and (5) Zip Ties.