Cole-Parmer® DC-400 Standard Desiccator Cabinets, Antylia Scientific
Supplier: Argos Technologies
Certificates
About this item
Safely store humidity sensitive products.
- Easily view samples through acrylic viewing panels
- Durable, rugged construction - Aluminum frame, acrylic panels, and stainless steel shelves
- Stainless steel shelves with holes ensure uniform humidity throughout chamber
Desiccators provide dust-free storage for humidity sensitive products such as optical equipment, electronic parts, PCB boards, and reagents. Place a desiccant material in the cabinet to keep the environment dry. Once the desiccant is saturated, simply replace or regenerate the desiccant for reuse. A lock and key allow you to securely lock the cabinet to prevent unauthorized access.
Delivery: Supplied with one stainless steel shelf and a key.
Specifications
- Catalog No:
- 76640-522
- 76640-520
- Capacity:
- 70 L
- Exterior dimensions:
- 50.2×38×58.1 cm (19.8×15×22.9")
- 43.3×40×43.3 cm (17×15.7×17")
- Int. volume:
- 70 L
- 50 L
Specifications
Customers Who Bought This Also Bought