Order Entry
Canada
ContactUsLinkComponent
SITVue /DAB Detection System, Diagnostic BioSystems
SITVue /DAB Detection System, Diagnostic BioSystems
Catalog # CA76635-216
CAS Number:  
SITVue /DAB Detection System, Diagnostic BioSystems
Catalog # CA76635-216
Supplier Number:  SIT-100D-RUO
CAS Number:  

Specifications

  • Description:
    SITVue/DAB detection system
  • Size:
    100 tests
  • Storage conditions:
    Keep away from light
  • Shelf life:
    12 - 36 months
  • Storage temperature:
    2...8 °C
  • Cat. no.:
    CA76635-216

Specifications

About this item

SITVue /DAB detection system is a powerful Intensification system that significantly enhances chromogenic signals. This system can be integrated into standard immunohistochemistry (IHC) staining methods.

SITVue /DAB Detection System is an enzyme-mediated reaction that utilizes horseradish peroxidase (HRP) to catalyze the deposition of two separate Linkers applied sequentially onto tissue sections or cell preparation. The deposited Linkers can be detected with a streptavidin-peroxidase conjugate followed by a reaction with a peroxidase substrate/chromogen solution such as diaminobenzidine (DAB).

SITVue /DAB Detection System results in a significant increase in sensitivity compared to standard IHC detection methods, while maintaining similar specificity.

Delivery: Kit contains: Tissue primer (10 ml), Linker 1 (10 ml), Linker 2 (10 ml), Tracer (10 ml), Stable DAB/Plus buffer (10 ml), Stable DAB/Plus chromogen (15 ml), Empty mixing bottle for Stable DAB/Plus (1 ml) and 15 ml bottle.