Cell mAb Medium, Quantum Yield, Gibco™ (a part of Thermo Fisher Scientific)
Catalog #: CA76628-924
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Description:Cell MAb medium, quantum yield, animal origin-free: Chemically defined
- Size:1000 ml
- Cat. no.:CA76628-924
Specifications
About this item
Cell MAb Medium, quantum yield, is a chemically defined (CD) basal cell culture medium. It contains L-glutamine and phenol red and does not contain Pluronic™ F68 (Kolliphor™ P188) or other surfactants.
- Chemically defined (CD)
- No pluronic acid and other surfactants
- Contains L-glutamine and phenol red
Cell MAb Medium, quantum yield, should be supplemented in the same way as traditional basal medium (e.g., serum or serum-free supplements).
Customers Who Bought This Also Bought