Cell mAb Medium, Animal Component Free, Gibco™ (a part of Thermo Fisher Scientific)
Catalog #: CA76628-942
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Description:Cell MAb medium, animal component free, animal origin-free: Chemically defined
- Size:1 L
- Cat. no.:CA76628-942
Specifications
About this item
Cell MAb Medium, animal component free, is a complete medium.
- Contains L-glutamine
- Supplemented with 0.3% Difco Select Soytone, and enzymatic digest of soy
- No phenol red, pluronic acid or other surfactants
It does not contain attachment factors, which may need to be added if this medium is used with adherent cell lines.
Customers Who Bought This Also Bought