ContactUsLinkComponent
Starter Pak No. 2, Gibco™ (a part of Thermo Fisher Scientific)
Starter Pak No. 2, Gibco™ (a part of Thermo Fisher Scientific)
Catalog # CA76628-730
CAS Number:  
Starter Pak No. 2, Gibco™ (a part of Thermo Fisher Scientific)
Catalog # CA76628-730
CAS Number:  
Specifications
  • Description:
    Starter Pak no. 2
  • Size:
    1 each
  • Cat. no.:
    CA76628-730
Specifications
About this item
Starter Pak No. 2 contain a variety of animal origin and animal origin-free peptones that offer many essential nutrients needed for the production of human and animal vaccines. It provides conveniently packaged, 100-g, non-GMP samples of these peptones that are best suited for vaccine production.
Provides conveniently packaged, 100-gram, non-GMP samples of these peptones that are best suited for vaccine production and contains:
Bacto Yeast Extract - this yeast has the highest level of carbohydrates of our yeast products and works wiell in a variety of human and animal health vaccines.
Phytone Peptone and Difco Soytone - both enzymatic digests of soy, these peptones are also a nutritious source of carbohydrates. These products work well in microbial fermentation processes, as well as in mammalian cell culture processes such as CHO. Blending these soy peptones with yeast peptones has been shown to provide additional benefits to cultures.
Bacto Proteose Peptone No. 2 and Bacto Proteose Peptone No. 3 - these enzymatic digests of porcine protein provide nutrition for fastidious microorganisms. Bacto Proteose Peptone No. 3 can replace serum in many applications and helps increase monoclonal antibody and recombinant protein product in CHO cells.
Bacto Casamino Acids - this supplement has low salt and iron content, making it an excellent supplement for media formulations for which nitrogen requirements are minimal.
The Starter Paks provide conveniently packaged samples of the peptones most commonly used in mammalian cell culture and microbial fermentation bioproduction processes. Starter Paks are tailored for specific applications, including the production of monoclonal antibodies, recombinant proteins, and vaccines.