Difco™ Brain Heart Infusion, without Dextrose, Gibco™ (a part of Thermo Fisher Scientific)
Catalog # CA76628-764
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Description:Difco™ Brain heart infusion, without dextrose, animal origin (Bovine, Porcine)
- Size:10 kg
- Cat. no.:CA76628-764
Specifications
About this item
Difco Brain heart infusion, without dextrose, is an animal origin (AO) enzymatic digest of bovine and porcine animal proteins.
- Contains the same formulation as Bacto Brain heart infusion with no dextrose added
- Basal medium that is generally used with carbohydrates for fermentation studies
- Can be used in tests for the verification of fecal Streptococci
Listed by the National Committee for Clinical Laboratory Standards (NCCLS) as a medium for use in the preparation of microdilution trays for antimicrobial susceptibility testing by the broth microdilution procedure.
Difco Brain heart infusion, without dextrose, is a microbiological culture medium used for cultivating fastidious microorganisms, including Streptococci, Pneumococci, and Meningococci.