CHO Medium, Gibco™ (a part of Thermo Fisher Scientific)
Catalog #: CA76628-902
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Description:CHO medium
- Size:10 kg
- Cat. no.:CA76628-902
Specifications
About this item
CHO Medium is a high-performance, complete, animal origin-free (AOF) formulation offering high-production yields without the need for further supplementation. This medium is formulated to support cell growth and production of antibodies and recombinant proteins in suspension CHO cell cultures.
- Chemically defined (CD)
- Protein-free
- Serum-free
CHO Medium can be used in various culture vessels in batch, fed-batch, and profusion processes for both research and manufacturing applications.
Customers Who Bought This Also Bought