Millennia™ 2000 Adhesion Slides
Supplier: StatLab
Certificates
About this item
Millennia™ 2000 slides feature a hydrophilic surface with strong adhesion properties.
- Hydrophilic slide designed for IHC, special stains, frozen sections, and routine histology
- Thermal transfer printer compatible
- Available with 45 degree clipped corners only
Part of the StatLab Millennia Adhesion Microscope Slide family. Manufactured in Japan.
Packaging: Slides are packaged 100 per box.
Frequently Bought Together