miniPCR® Agarose Tabs™
Agarose in convenient tablet format - no weighing required!
- For 2% agarose gels: Mix 1 tab with 25 ml 1X TBE buffer (to yield 1 to 2 agarose gels for blueGel™ system)
- For 1% agarose gels: Mix 1 tab with 50 ml 1X TBE buffer (to yield 3 agarose gels for blueGel™ system)
Electrophoresis-grade agarose in a convenient tablet format - no messy weighing required!
Pre-weighed tablets of standard melting point agarose in a convenient blister pack, Agarose Tabs™ deliver high resolution and sharp bands with high transparency and low background. They enhance ease of gel pouring and handling in a lab or classroom. Agarose Tabs™ are ideal for routine DNA and RNA gel electrophoresis including separation of PCR products.
With 0.5 g agarose per tab, one pack is sufficient to cast up to 30 1% gels or up to 15 2% gels for the blueGel™ electrophoresis system.
Frequently Bought Together