miniPCR® SeeGreen™ All-in-One Agarose Tabs™
Supplier: AMPLYUS LLC
Certificates
About this item
All-in-one tablets with pre-measured agarose, TBE, and SeeGreen™ stain.
- All-in-one tabs: Agarose, SeeGreen™ stain, TBE buffer
- Cast 20 gels (2%) to 40 gels (1%) per box
- Ideal for DNA and RNA visualization with blue light or UV
- Fast dissolving-gels ready to pour in seconds without weighing or mixing
- High transparency, low background, high sensitivity (0.2 ng DNA or 1 ng total RNA)
- Stable at room temperature for >2 years
Add water, boil, and pour!
All-in-one tabs with pre-measured agarose, SeeGreen™ nucleic acid stain, and the TBE buffer needed to cast electrophoresis gels. SeeGreen™ All-in-One Agarose Tabs™ contain SeeGreen™ stain to facilitate DNA or RNA visualization under safe blue light, making them ideal for use with the blueGel™, GELATO™, or any electrophoresis system.
Packaged in a convenient blister pack, these tabs are ideal for routine DNA and RNA gel electrophoresis including separation of PCR products. With 0.4 g agarose per tab, one box is sufficient to cast 40 1% gels or 20 2% gels for the blueGel™ system (20 ml). Stable at room temperature for 2 years.
Frequently Bought Together