Inoculating Loops
Inoculating loops
Catalog # 470334-904
Supplier: GSC International
Supplier #: 4-23515
Jump to:
Product Details & Documents
A necessary tool for working with bacteria.
- Looped nichrome wire
- Attached metal handle
These simple tools are ideal for retrieving an inoculum from a culture or transferring microorganisms to streak plates or cultures. This needle is made out of nichrome wire and has a loop at the end. It is attached to a 6" long metal handle.
Specifications
- Description:Inoculating loops