COSA™ Foam 40, Ecolab® Life Sciences
76637-528
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:COSA™ Foam 40
- Size:1 gal.
- Environmentally Preferable:
- Cat. no.:76637-528
COSA™ Foam 40 is a liquid, neutral pH product for manual cleaning, or for use as a foam additive in cleaning solutions with Cosa™ CIP products for the pharmaceutical and personal care industries.
- Suitable for manual and foam cleaning
- Clear solution at all temperatures
- Low toxicity high LD50-value and PDE value
- cGMP manufactured - Global formulation
Safe for manual use, COSA™ Foam 40 acts as a mild and odorless detergent.