ContactUsLinkComponent
Innovating Science® Wide Range pH Indicator Kit
Catalog # 470348-616
Supplier:  Ward's Science
undefined
Innovating Science® Wide Range pH Indicator Kit
Catalog # 470348-616
Supplier:  Ward's Science
Supplier Number:  IS5016
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Wide range pH water test kit
  • Environmentally Preferable:
  • Cat. no.:
    470348-616
  • Includes:
    Instructions and SDS
  • No. of Uses:
    50
Specifications
About this item
Easy way to test for pH of water within 2 to 10 range.
  • Over 50 tests to measure pH of water samples
Test water pH within 2 to 10 range with this wide range pH testing kit. Kit includes enough materials to repeat the procedure over 50 times.