CompactDry™ Pseudomonas aeruginosa (PA), Hardy Diagnostics
Catalog # CA76628-082
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Description:CompactDry™ Pseudomonas aeruginosa (PA) cassette, for enumeration of P. aeruginosa in foods, beverages, and water
- Size:25 packs of 4/100 plates per pack
- Cat. no.:CA76628-082
Specifications
About this item
CompactDry™ Pseudomonas aeruginosa (PA) is a ready-to-use test method recommended for the enumeration of Pseudomonas aeruginosa in foods, beverages, water, cannabis, and more.
- Sturdy frame, stackable packaging
- Self-diffusing media
- Ability to subculture without damage to colonies
- No leakage
- Easy to label
CompactDry™ PA is suitable for testing a broad range of water types for human consumption or other related industries.
Each cassette comes with four connected plates that can be easily snapped apart to support a variety of dilution scenarios.
Caution: Stored at room temperature vs the competitor products with storage at temperatures less than or equal to 8 °C.