ContactUsLinkComponent
EnzyFluo™ Human/Mouse Rps6(S235/236) Phosphorylation ELISA, BioAssay Systems
  76545-508
 :  
undefined
EnzyFluo™ Human/Mouse Rps6(S235/236) Phosphorylation ELISA, BioAssay Systems
  76545-508
 :  ERPS6-100
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Size:
    100 tests
  • Cat. no.:
    76545-508
Determination of RPS6(S235/236) phosphorylation status in whole cells and evaluation of pathway modulation by activators, inhibitors, siRNA etc.
  • New and improved: Total assay time reduced from the standard 9 to 5 hours
  • Simple and convenient: Cells are directly cultured in 96-well plates, no cell lysis necessary
  • Accurate: Protein phosphorylation is normalized to total cellular protein in the same well
  • Method: Cell-Based ELISA (FL530/585 nm, 360/450 nm)
  • Samples: Cells
  • Species: human, rat and mouse
PROTEIN PHOSPHORYLATION plays key roles in regulating protein activity within cells. A large number of protein kinases and phosphatases are known to be involved in a variety of signal transduction pathways. A rapid assay capable of monitoring such signal transduction pathways, target protein phosphorylation status and drug effects is highly desirable in life science research and drug discovery.
Ribosomal Protein S6 (RPS6) is one of the ribosomal proteins of the 40S subunit of eukaryotic ribosomes. RPS6 plays important roles in biosynthesis of cell's translational apparatus, a critical component for cell growth and proliferation. BioAssay Systems' RPS6(S235/236) Phosphorylation ELISA Kit provides a rapid and sensitive assay for measuring the phosphorylated protein pRPS6(S235/236) in cultured cells and normalizes the level of phosphorylation to the total protein content in the same well.
This simple and efficient assay eliminates the need for cell lysate preparation and can be used to study the underlying signal pathway and effects of inhibitors, siRNA or activators on the pathway. In this assay, cells grown in a 96-well plate are fixed and permeabilized. The phosphorylation of RPS6(S235/236) is measured using a specific primary antibody followed by a HRP-conjugated secondary antibody with a fluorogenic substrate, and a fluorescent reagent that measures the total protein in the same well.