QuantiChrom™ HRP Detection Reagent, BioAssay Systems
76545-500
:
- Size:200 tests
- Cat. no.:76545-500
Direct assays of peroxidases, ELISA, immunochromogenic chemistry, western blot.
- Method: OD 450 nm
- Samples: Peroxidases and HRP in ELISA
- Species: All
- Detection limit: 76 nU/well, or 7.7 pg/well
HORSERADISH PEROXIDASE (HRP) is an enzyme commonly used when conjugated to analytes of interest. When combined with hydrogen peroxide and a dye, HRP can be used to colorimetrically quantify the analyte.