Orders LinkContactUsLinkComponent
Stopcocks, plain bore, PYREX® Rotaflo®
Stopcocks, plain bore, PYREX® Rotaflo®
Supplier:   Corning
Stopcocks, plain bore, PYREX® Rotaflo®
Supplier:  Corning
Certificates
Stopcocks, plain bore, PYREX® Rotaflo®
Stopcocks, plain bore, PYREX® Rotaflo®
8 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass.
  • Fine flow adjustment
  • Lubrication is not required
  • Fast assembly
These high performance Rotaflo® stopcocks can be adjusted with bore openings and have sidearms. Only chemically resistant PTFE and PYREX® borosilicate glass are in contact with liquids and gase. Seal is visible when open or closed.
Documents
Product Family Options
Two-way, in-line pattern
Product Information
  • Sidearm Dimensions (Ø×L)
    Bore size
    Availability
    Price
  • 8×210 mm
    0 - 3 mm (0 - 0.118")
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $248.00
    $248.00
    Case of 1
       $248.00
    Supplier Number: 7470-3
  • 10×210 mm
    0 - 6 mm (0 - 0.236")
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $331.00
    $331.00
    Case of 1
       $331.00
    Supplier Number: 7470-6
  • 13×224 mm
    0 - 10 mm (0 - 0.394")
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $381.00
    $381.00
    Case of 1
       $381.00
    Supplier Number: 7470-10
Two-way, right angle pattern
Product Information
  • Sidearm Dimensions (Ø×L)
    Bore size
    Availability
    Price
  • 8×155 mm
    0 - 3 mm (0 - 0.118")
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $235.00
    $235.00
    Case of 1
       $235.00
    Supplier Number: 7473-3
  • 10×160 mm
    0 - 6 mm (0 - 0.236")
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $345.00
    $345.00
    Case of 1
       $345.00
    Supplier Number: 7473-6
  • 13×175 mm
    0 - 10 mm (0 - 0.394")
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $393.00
    $393.00
    Case of 1
       $393.00
    Supplier Number: 7473-10
Three-way
Product Information
  • Sidearm Dimensions (Ø×L)
    Bore size
    Availability
    Price
  • 8×215 mm
    0 - 3 mm (0 - 0.118")
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $403.00
    $403.00
    Case of 1
       $403.00
    Supplier Number: 7475-3
  • 10×220 mm
    0 - 6 mm (0 - 0.236")
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $416.00
    $416.00
    Case of 1
       $416.00
    Supplier Number: 7475-6