Corning® Cryogenic Vial Grippers, Corning
Catalog #: 75779-848
Supplier: Corning
Specifications
- Description:Cryogenic Vial Grippers, Pack of 5, Assorted Colors
- Cat. no.:75779-848
- Supplier No.:432136
Specifications
About this item
These cryogenic vial grippers feature a unique design to grasp internal or external-thread cryogenic vials.
They enable to easily sort or move vials while maintaining sterility and protecting fingers from frozen vials, dry ice and liquid nitrogen.
Customers Who Bought This Also Bought