Orders LinkContactUsLinkComponent
Corning® Internal Thread Cryogenic Vials, Corning
Corning® Internal Thread Cryogenic Vials, Corning
 :   Corning
Corning® Internal Thread Cryogenic Vials, Corning
 :  Corning
Corning® Internal Thread Cryogenic Vials, Corning
Corning® Internal Thread Cryogenic Vials, Corning
These vials are manufactured from polypropylene to withstand temperatures down to –196 °C.
  • Black graduations with large marking spot
  • Vials have a silicone washer for a secure seal
  • 2 ml self-standing vials are available in five cap colors.
  • Vials may be color coded with inserts
  • Free foam rack with each case
  • RNase-/DNase-free
  • Sterile
  • Nonpyrogenic
Self-standing vials have a special base design allowing them to be locked into cryogenic rack and tray (Cat. No. 431131) for single-handed manipulation.
 : Do not use cryogenic vials for storage in the liquid phase of liquid nitrogen. Only store vials in the vapor phase above the liquefied gas. Always use appropriate safety equipment when removing vials from cryogenic storage.
  • Catalog No:
  • 66021-940
  • 66021-944
  • 66021-946
  • 66021-950
  • 89089-764
  • 66021-942
  • 66021-948
  • 66021-970
  • 30617-556
  • 30617-558
  • 30617-560
  • 30617-562
  • 30617-564
  • 30617-566
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
Product Family Options
  • Capacity
    Bottom style
    Cap included
    Cap color
  • 1.20 ml
    Conical bottom, self-standing
    Yes
    Orange
    Case of 500
    Case of 500 - $457.30
    $457.30
    Case of 500
       $457.30
     : 430487
      Low Manufacturing Impact
    Specifications
    Cat. No.
    66021-940
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom
    Yes
    Orange
    Case of 500
    Case of 500 - $650.25
    $650.25
    Case of 500
       $650.25
     : 430489
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    66021-944
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 4.00 ml
    Round bottom
    Yes
    Orange
    Case of 500
    Case of 500 - $639.20
    $639.20
    Case of 500
       $639.20
     : 430490
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    66021-946
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 5.00 ml
    Round bottom
    Yes
    Orange
    Case of 500
    Case of 500 - $746.30
    $746.30
    Case of 500
       $746.30
     : 430492
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    66021-950
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom, self-standing
    Yes
    Orange
    Case of 250
    Case of 250 - $279.48
    $279.48
    Case of 250
       $279.48
     : 431386
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89089-764
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom, self-standing
    Yes
    Orange
    Case of 500
    Case of 500 - $543.00
    $543.00
    Case of 500
       $543.00
     : 430488
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    66021-942
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 4.00 ml
    Round bottom, self-standing
    Yes
    Orange
    Case of 500
    Case of 500 - $652.81
    $652.81
    Case of 500
       $652.81
     : 430491
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    66021-948
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 5.00 ml
    Round bottom, self-standing
    Yes
    Orange
    Case of 500
    Case of 500 - $746.30
    $746.30
    Case of 500
       $746.30
     : 430656
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    66021-970
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom, self-standing
    Yes
    Assorted
    Case of 500
    Case of 500 - $573.75
    $573.75
    Case of 500
       $573.75
     : 431416
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    30617-556
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom, self-standing
    Yes
    Yellow
    Case of 500
    Case of 500 - $521.05
    $521.05
    Case of 500
       $521.05
     : 431417
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    30617-558
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom, self-standing
    Yes
    Blue
    Case of 500
    Case of 500 - $521.05
    $521.05
    Case of 500
       $521.05
     : 431418
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    30617-560
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom, self-standing
    Yes
    Green
    Case of 500
    Case of 500 - $521.05
    $521.05
    Case of 500
       $521.05
     : 431419
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    30617-562
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom, self-standing
    Yes
    Red
    Case of 500
    Case of 500 - $521.05
    $521.05
    Case of 500
       $521.05
     : 431420
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    30617-564
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2.00 ml
    Round bottom, self-standing
    Yes
    White
    Case of 500
    Case of 500 - $521.05
    $521.05
    Case of 500
       $521.05
     : 431421
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    30617-566
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
Cap inserts
  •  
    Color
    Description
  • White
    Color-Coded Cap Inserts
    Case of 500
    Case of 500 - $95.20
    $95.20
    Case of 500
       $95.20
  •  
    Color
    Description
  • Blue
    Color-Coded Cap Inserts
    Case of 500
    Case of 500 - $95.20
    $95.20
    Case of 500
       $95.20
  •  
    Color
    Description
  • Red
    Color-Coded Cap Inserts
    Case of 500
    Case of 500 - $95.20
    $95.20
    Case of 500
       $95.20
  •  
    Color
    Description
  • Green
    Color-Coded Cap Inserts
    Case of 500
    Case of 500 - $95.20
    $95.20
    Case of 500
       $95.20
  •  
    Color
    Description
  • Yellow
    Color-Coded Cap Inserts
    Case of 500
    Case of 500 - $95.20
    $95.20
    Case of 500
       $95.20
  •  
    Color
    Description
  • Assorted
    Color-Coded Cap Inserts
    Case of 500
    Case of 500 - $92.65
    $92.65
    Case of 500
       $92.65