Order Entry
Canada
ContactUsLinkComponent
 
Eisco Plastic Narrow Neck Conical Flasks
  470345-174
 :  Wards
undefined
Eisco Plastic Narrow Neck Conical Flasks
  470345-174
 :  Wards
 :  CH0431G

 

  • Capacity:
    250 ml
  • Color:
    Translucent
  • Cat. no.:
    470345-174

 

 

Polypropylene conical flasks with narrow neck and no graduations.

  • Made from high-quality polypropylene
  • Highly chemical resistant, microwavable and autoclavable at 121 °C
  • Narrow neck and conical shape prevents spills
  • Translucent body makes it easy to see liquid level in the flask

Conical flasks with a narrow neck and natural translucent color, making it easy to see any liquid level in the bottle. Made from easy-to-clean high-quality polypropylene. Highly chemical resistant, microwavable and autoclavable at 121 °C.