Specifications
- Sterility:Sterile
- Color:Clear
- Description:Deep well plates
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Cat. no.:47745-406
- Packaging:Individually wrapped
- Working Well Volume:240 µl
- Supplier No.:P384240SQCSI
Specifications
About this item
PP. Used for sample collection, cell culture and long-term storage.
- Square wells
- Excellent chemical resistance
- Temperature resistant down to −80 °C
- Can withstand centrifugation up to 4000 ×g
Two types of sealing mats are suitable for deep 384-well microplates:
AxyMat™ sealing mats are designed for high throughput screening and storage applications. Made of silicone, they can be autoclaved, and following a bleach wash and ethanol rinse protocol, they can also be re-used. AxyMat™ sealing mats are pierceable and self-sealing, ideal for automated processes, and can be used in sub-zero environments.
ImpermaMat™ sealing mats are chemically resistant. They are made for use in assays involving strong solvents such as DMSO and similarly harsh solvents. Re-usable. They feature a tight seal to the plate so there is no sample evaporation or well-to-well evaporation. Usable within a wide temperature range: from −80 up to +121 °C.