PA43 BIO Trinocular Ergonomic Upright Pathology Lab Microscope
Catalog # CA76615-054
Supplier: Motic
New Product
Specifications
- Description:PA43 BIOMED microscope
- Cat. no.:CA76615-054
Specifications
About this item
The new PA43 BIOMED is a new series from Motic scientific and represents a large step forward in combining modularity with ergonomics to improve application imaging workflows. A progression into routine work, this new microscope’s robust design and solid footprint does not compromise on optical quality and features.
- Kohler illumination with option of halogen or LED illumination
- Environmentally friendly ECO function
- Consistent, even illumination with IL function
- Rack less stage
Its modularity offers flexibility when choosing a configuration that suits multiple end-user applications from cost-effective solutions to high-quality optical systems for lab diagnostics. The microscope is not only modular but also flexible in design and can be upgraded at any time.
Ordering information: Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.