Trinocular Research Upright Trinocular with LUMOS EPI Flourescence LED, PA53 BIO
Catalog # CA76614-944
Supplier: Motic
New Product
Specifications
- Description:PA53 FS6 with LUMOS FL LED
- Cat. no.:CA76614-944
Specifications
About this item
The PA53 BIO FS6 microscopes offer maximum flexibility for managing any kind of fluorescence application. The high-power 3-channel LUMOS LED or the traditional mercury lamp, is up to the customer to select the desired excitation source.
- Coded fluorescence turret automatically remembers the fluorescence channel if changed
- Motic fluorescence imaging software plug-in for Motic imaging plus 3.0 is included with all PA53 BIO FS6 packages
- Perfect light source for fluorescence: LUMOS FL
The fluorescence filter cubes are mounted in an encoded six-position turret: Multi-color staining can easily be handled. The standard configuration of PA53 BIO FS6 with DAPI, FITC, and TRITC filter cubes is open for more filters and more demands. A multiband filter (DAPI/Green/Orange) is an available as an option for FISH technique. Our H-Plan S-APO (fluor) objectives provide high transmission rates for the brightest images. No need for Plan Apos: Too much glass inside.
Ordering information: Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.