Corning® BioCoat® Poly-D-Lysine/Laminin German Glass Coverslip, 12 mm, Corning
Catalog #: CACB354087
Supplier: Corning
Specifications
- Description:12 mm, round coverslips with poly-D-lysine/laminin
- Cat. no.:CACB354087
- Supplier No.:354087
Specifications
About this item
Corning® BioCoat™ Poly-D-Lysine/Laminin 12mm #1 German Glass coverslip with a uniform application of poly-D-lysine and laminin. Bulk packed, 80/case. Corning BioCoat products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
- Shelf life: >= 3 months from date of shipment
Customers Who Bought This Also Bought