Corning® BioCoat® Fibronectin German Glass Coverslip, 22 mm, Corning
Catalog #: CACB354088
Supplier: Corning
Specifications
- Description:Round, 22 mm
- Cat. no.:CACB354088
- Supplier No.:354088
Specifications
About this item
Corning® BioCoat™ 22×22 mm #1 German Glass coverslip with a uniform application of human fibronectin. Bulk packed, 60/case. Corning BioCoat products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
- Shelf life: >= 3 months from date of shipment
Customers Who Bought This Also Bought