Corning® BioCoat® Poly-L-Lysine, German Glass Coverslip, 12 mm, Corning
Catalog #: CACB354085
Supplier: Corning
Specifications
- Description:Round coverslips with Poly-L-Lysine, 12 mm
- Cat. no.:CACB354085
- Supplier No.:354085
Specifications
About this item
Corning® BioCoat™ 12 mm #1 German Glass coverslip with a uniform application of Poly-L-Lysine. 40/pack, 80/case. Corning BioCoat products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
- Shelf life: >= 3 months from date of shipment
Customers Who Bought This Also Bought