Corning® BioCoat® Collagen I 96-well White/Clear Flat Bottom TC-treated Microplate with Lid, Corning
Catalog # CA62406-386
Supplier: Corning
Specifications
- Well Volume:392 µl
- Color:White/Clear
- Description:96-well flat bottom TC-treated microplate, with lid
- Cat. no.:CA62406-386
- Plate Color:Clear bottom white
- Sterile:Non sterile
- Supplier No.:356650
Specifications
About this item
Corning® BioCoat™ Collagen I 96-well white/clear microplate is comprised of white TC-treated polystyrene with a uniform application of rat tail collagen type I.
- Clear flat bottom, with lid